Learn More
MSC Miller TurboLite+ Scorpion Personal Fall Limiter, 9 ft., 420 lb., 1 ea.
Supplier: MSC 78089372
This self-retracting fall limiter is a personal fall protection device designed for use in construction, maintenance, and other high-risk environments. It features a 9 ft. (2.7 m) lifeline made of abrasion-resistant webbing with a Vectran core and polyester jacket, providing greater freedom of movement compared to traditional shock-absorbing lanyards. The compact design is suitable for confined spaces and includes a visual load indicator to alert users when the unit requires removal from service.
- Quick-activating stainless steel braking mechanism
- Steel locking snap hook lifeline connector
- High-impact resistant nylon housing
- Meets OSHA 1926.502 and 1910.140, ANSI Z359.14-2021 Class 1, and CSA Z259.2.2-17 Class SRL standards
- No annual recertification required
- Maximum capacity of 420 lb. (per OSHA standards)