Learn More
MSC Scot's Tuff Cleaner, 32 oz, Citrus Scent, 1 ea
Catalog No. 502866109
Encompass
Scot's Tuff liquid toilet bowl cleaner is a non-acid, ready-to-use disinfectant solution designed for effective cleaning and disinfection of hard nonporous surfaces.
  • Formulated as a liquid
  • Packaged in a convenient 32 oz bottle
  • Features a pleasant citrus fragrance
  • Suitable for use on toilet bowls, urinals, and other hard surfaces
  • Maintains a pH range of 11.5–12.5
Catalog No. 50-286-6109 Supplier MSC Supplier No. 09309071
Scot's Tuff liquid toilet bowl cleaner is a non-acid, ready-to-use disinfectant solution designed for effective cleaning and disinfection of hard nonporous surfaces.
  • Formulated as a liquid
  • Packaged in a convenient 32 oz bottle
  • Features a pleasant citrus fragrance
  • Suitable for use on toilet bowls, urinals, and other hard surfaces
  • Maintains a pH range of 11.5–12.5