Learn More
Paul N Gardner Company SCOT VOLUMETER PAINT PIG 40MES
SDP
Catalog No. NC0087856
Encompass
The Paint-Pigment Volumeter is used to determine apparent density of dry pigments or other free flowing, fine powders. It provides a means of reproducibly packing a tared cubic inch measure and then weighing the filled container.
Catalog No. NC0087856 Supplier Paul N Gardner Company Supplier No. WG0601
The Paint-Pigment Volumeter is used to determine apparent density of dry pigments or other free flowing, fine powders. It provides a means of reproducibly packing a tared cubic inch measure and then weighing the filled container.