Learn More
Electron Microscopy Sciences Scotch™ 665 Tape, 3/4" no liner 36 YD
SDP
Supplier:  Electron Microscopy Sciences 77102
Encompass_Preferred
Non-conductive transparent tape with adhesive on both sides. Without is no liner to remove. Long lasting for permanent applications. 3"core.
Catalog No. 50-285-27