Learn More
Electron Microscopy Sciences Scotch™ 665 Tape, 3/8" no liner 72 YD
SDP
Catalog No. 5028526
Encompass_Preferred
Non-conductive transparent tape with adhesive on both sides. Without is no liner to remove. Long lasting for permanent applications. 3"core.
Catalog No. 50-285-26 Supplier Electron Microscopy Sciences Supplier No. 77101
Non-conductive transparent tape with adhesive on both sides. Without is no liner to remove. Long lasting for permanent applications. 3"core.