Learn More
RS Hughes Inc 3M Scotch 800 Clear Label Protection Tape
SDP
Catalog No. NC0264902
Encompass_Preferred
3M Scotch 800 clear label protection tape uses an acrylic adhesive and is packaged 36 rolls per case. In addition to the other properties this product is abrasion-resistant, moisture-resistant, UV-resistant.
Catalog No. NC0264902 Supplier RS Hughes Inc Supplier No. 02120003551RL
3M Scotch 800 clear label protection tape uses an acrylic adhesive and is packaged 36 rolls per case. In addition to the other properties this product is abrasion-resistant, moisture-resistant, UV-resistant.