Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Bi-Directional Filament Tape

Catalog No. 19072002 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
50 m x 50 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-072-002 50 m x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-002 Supplier 3M™ Supplier No. 7000096086
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Scotch™ Bi-Directional Filament Tape is a high strength, general purpose, clear fiberglass-reinforced tape with a synthetic rubber resin adhesive ideal for heavy box closure and cross web splicing.

  • Heavy-duty, cross directional, high strength tape for bundling, reinforcing and pallet unitizing applications
  • Resists center-seam splitting in RSC box closure applications.
  • Bi-directionally fiberglass reinforced tape provides cut and tear resistance in machine direction and cross direction
  • Synthetic rubber adhesive bonds easily to most surfaces with high initial adhesion
  • Transparency provides clear visibility of box graphics, printing and illustrations
  • Able to withstand rough shipping and handling conditions
TRUSTED_SUSTAINABILITY

Specifications

Product Type Filament Tape
For Use With (Application) Heavy Duty Box Sealing
Adhesive Type Synthetic Rubber
Color Clear
Dimensions (L x W) 50 m x 50 mm
Material Synthetic Rubber
Model 8959
Quantity 18 Rolls/Cs.
Thickness (English) 5.7 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.