Learn More
Staples Advantage Scotch™ Brand Transparent Tape
Description
- Keeps labels clean and clearly readable
- Can be used for splicing, mending, laminating
- Dimensionally stable up to 107°C (225°F)
- Resists degradation due to sunlight exposure, water, extended storage
- Matte-finish acetate tape disappears when applied, stay invisible over time, and can be written on
- Gloss-finish acetate tape is stronger than matte finish tapes; its 2 in. (51mm) width makes it perfect for covering large labels
Order Info
Tapes with 3 in. core fit 11-865-6 dispenser.
Specifications
Specifications
| Dimensions (L x W) | 108 ft. x 0.75 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.