Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch-Brite™ Easy Erasing Pad

Catalog No. 17623918 Shop All 3M Products
Change view
Click to view available options
Color:
Blue, White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-918 Blue, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-918 Supplier 3M™ Supplier No. 7100032938
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Scotch-Brite™ Easy Erasing Pad is ideal for removing stains, marks, ink and more. Just use water - no chemicals or sprays needed.

  • Soft, flexible pad cleans many surfaces without using additional chemicals - just use water
  • Tough on hard to remove stains
  • Blue urethane foam sponge holds water to keep pad moist, can be used to wipe after use, and keeps the pad intact over its useful life
TRUSTED_SUSTAINABILITY

Specifications

Product Type Scrub Sponge
Color Blue, White
For Use With (Application) General Surface Cleaning
Length (English) 4.5 in.
Length (Metric) 114 mm
Material Polymeric Foam
Model 4004 CC
Quantity 12/Cs.
Width (English) 2.8 in.
Width (Metric) 71 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.