Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch-Brite™ Extra Heavy Duty Pot 'N Pan Scour Pad

Catalog No. 17623900 Shop All 3M Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-900 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-900 Supplier 3M™ Supplier No. 7010029029
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M Extra heavy duty scouring pad designed for removing stubborn baked on cheeses and sauces from cooking pots, pans, utensils and cooking equipment. Thick, open construction allows easy rinsing. Avoids the risk of splintering and rusting like a stainless steel scrubber, while getting the job done.

  • Thick, open mesh construction.
  • Rinses out easily, doesn’t splinter and won’t rust.
  • Can replace stainless steel scrubbers
  • Certified by HACCP International as 'Food Safe' and 'Fit for Purpose.'
TRUSTED_SUSTAINABILITY

Specifications

Product Type Scouring Pad
Certifications/Compliance HACCP International
Color Blue
For Use With (Application) Commercial Kitchen Cleaning
Length (English) 5 in.
Length (Metric) 127 mm
Material Fiber, Mineral, Resin
Quantity 40/Cs.
Width (English) 3.5 in.
Width (Metric) 89 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.