Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch-Brite™ General Purpose Scrub Pad

Catalog No. 17623865 Shop All 3M Products
Change view
Click to view available options
Color:
Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-865 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-865 Supplier 3M™ Supplier No. 7000052522

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M™ Medium duty scouring pad designed for everyday cleaning. Thicker, hand sized version of Scotch Brite™ General Purpose Scouring Pad 96. The convenient size eliminates the need to cut or fold the pad.

  • Thicker, hand-sized version of Scotch-Brite™ General Purpose Scouring Pad 96.
  • Cleans pots, pans and kitchen equipment
  • Replaces steel wool and metal sponges
  • Certified by HACCP International as 'Food Safe' and 'Fit for Purpose.'
TRUSTED_SUSTAINABILITY

Specifications

Product Type Scouring Pads
Certifications/Compliance HACCP International
Color Green
For Use With (Application) Commercial Kitchen Cleaning
Length (English) 4.5 in.
Length (Metric) 114 mm
Material Fiber, Mineral, Resin
Model 9650
Quantity 80/Cs.
Width (English) 3 in.
Width (Metric) 76 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.