Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch-Brite™ Power Pad 2000

Catalog No. 17623839 Shop All 3M Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-839 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-839 Supplier 3M™ Supplier No. 7000029659
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Scotch-Brite™ Power Pad 2000 scours 2 to 4 times faster than conventional commercial scouring pads, and is 6 to 10 times less abrasive than other medium/heavy duty commercial scouring products to reduce scratching. Unique bow tie shape.

  • Unique bow-tie shape for ease of handling
  • Scours at least 7 times faster than competitive non-scratch pads
  • 6 to 10 times less abrasive than other conventional commercial scouring pads
  • Certified by HACCP International as 'Food Safe' and 'Fit for Purpose.'
  • High quality bonding resin system designed to provide resistance to degradation from exposure to hot water, detergents and normal cleaning liquids
TRUSTED_SUSTAINABILITY

Specifications

Product Type Scouring Pad
Certifications/Compliance HACCP International
Color Blue
For Use With (Application) Commercial Kitchen Cleaning
Length (English) 5.5 in.
Length (Metric) 140 mm
Material Fiber, Mineral, Resin
Quantity 20/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.