Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch-Brite™ Stainless Steel Scrubber

Catalog No. 17623933 Shop All 3M Products
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-933 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-933 Supplier 3M™ Supplier No. 7100083743
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Scotch-Brite™ High quality stainless steel scrubber for tough cleaning chores. Removes baked on food from pots, pans, baking dishes, grills and ovens. Two sizes.

  • Single wire strand construction to eliminate multiple ends
  • High quality grade stainless steel to withstand breakage under normal usage and provide durability and corrosion resistance from exposure to hot water, detergents and normal cleaning liquids
  • Use for cleaning, scouring, and scrubbing pots, pans, baking dishes, broiler pans, grills and ovens
TRUSTED_SUSTAINABILITY

Specifications

Product Type Metal Scrubber
Color Gray
For Use With (Application) Pot and Pan Cleaning
Material Stainless Steel
Quantity 12/Cs.
Volume (English) 1.75 oz.
Volume (Metric) 49.6 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.