Learn More
Description
- Requires only neutral cleaner or water to effectively remove floor finish, preparing the floor for finish application
- Aggressively scrubs finishes in preparation for recoating
- Use with rotary or automatic floor machines
- Heavy duty scrubbing for 2 to 4 coat removal prior to recoating
- Helps reduce labor hours and material costs related to stripping floors
- Helps enhance safety and productivity: no slippery residue from stripper; no cleanup on baseboards or nearby floors; no chemical contamination on skin, clothes and shoes
- *Certified by Green Seal™ for Environmental Innovation based on a design for significantly improved mid-life performance compared to competitive products, extending the useful life of the floor pad and reducing solid waste generation
Specifications
Specifications
| Product Type | Stripping Pads |
| Color | Maroon |
| Quantity | 10 |
| For Use With (Application) | Scrubbing |
| Material | Non-woven Fiber |
| Diameter (Metric) | 355 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.