Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Filament Tape

Catalog No. 19072117 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
55 m x 24 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-072-117 55 m x 24 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-117 Supplier 3M™ Supplier No. 7000123442
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Scotch™ Filament Tape is a medium strength, general purpose, clear fiberglass-reinforced tape -with a synthetic rubber resin adhesive ideal for medium-duty strapping, bundling and reinforcing

  • Medium-duty, high strength tape for utility bundling, reinforcing and pallet unitizing applications
  • Synthetic rubber adhesive bonds easily to most fiberboard, plastics and metal surfaces
  • Glass yarn filaments add strength for box closing, bundling and more
  • Tape resists nicks, abrasions, moisture and scuffing
  • Transparency provides clear visibility of box graphics, printing and illustrations
TRUSTED_SUSTAINABILITY

Specifications

Product Type Industrial Tape
For Use With (Application) Box Closing, Bundling, Palletizing, Reinforcement, Strapping
Adhesive Type Synthetic Runner
Color Clear
Dimensions (L x W) 55 m x 24 mm
Material Synthetic Rubber
Model 897
Quantity 36 Rolls/Cs.
Thickness (English) 5 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.