Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Medium Performance Packing Tape

Catalog No. 19072082 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
50 m x 48 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-072-082 50 m x 48 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-082 Supplier 3M™ Supplier No. 7000042511

missing translation for 'validMainframeMsg'

Scotch™ Box Sealing Tape 372 is a medium performance packing tape that securely closes a wide variety of medium weight box materials, including recycled fiberboard.

  • Medium performance packaging tape for medium-duty applications on a wide range of materials
  • High edge tear and split resistance ideal for palletized and unitized loads
  • Polypropylene backing with synthetic rubber adhesive delivers conformability under stress
  • Easy unwind and smooth dispensing works well on high-speed automated packaging lines
  • Resists abrasion, moisture, chemicals and scuffing for long-lasting performance
  • Excellent adhesion to recycled liner paper and fiberboard
  • Polypropylene film backing is easy to unwind and resists abrasion, moisture and scuffing.
  • Backing easily conforms around edges and on rough surfaces for a tight seal.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Box Sealing Tape
For Use With (Application) Automated Carton Sealing, Box Closure, Branding
Adhesive Type Hot Melt Synthetic Rubber Resin
Color Clear
Description Medium performance
Dimensions (L x W) 50 m x 48 mm
Material Hot Melt Synthetic Rubber Resin
Model 372
Quantity 36/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.