Learn More
Electron Microscopy Sciences Scotch™ Photographic Tape 1/2"
SDP
Supplier:  Electron Microscopy Sciences 77400
Encompass_Preferred
Black, crepe paper, pressure-sensitive tape that is designed for use in commercial as well as private photography. It can be used for making negatives, positives, and transparencies. Conformable, opaque, and it has low light reflectance. 3" core. 3" core.
Catalog No. 50-285-06