Learn More
Electron Microscopy Sciences Scotch™ Photographic Tape, 1"
SDP
Catalog No. 5028505
Encompass_Preferred
Black, crepe paper, pressure-sensitive tape that is designed for use in commercial as well as private photography. It can be used for making negatives, positives, and transparencies. Conformable, opaque, and it has low light reflectance. 3" core. 3" core.
Catalog No. 50-285-05 Supplier Electron Microscopy Sciences Supplier No. 77410
Black, crepe paper, pressure-sensitive tape that is designed for use in commercial as well as private photography. It can be used for making negatives, positives, and transparencies. Conformable, opaque, and it has low light reflectance. 3" core. 3" core.