Learn More
Description
- Low odor and non-flammable properties for a safer working environment compared to typical acrylic adhesives
- Durable bond with excellent shear, peel and high impact strength
- 18-month room temperature shelf life simplifies inventory management
- Features spacer beads to control bond line thickness
- 10 minute work life with 21 minutes to structural strength
- Non-sag, toughened, two-part acrylic adhesive.
- Reaches structural strength twice as fast as most other competitive acrylic adhesives with a comparable open time
- 3M Catalog# 8810NS
Specifications
Specifications
| Adhesive Type | Acrylic |
| Color | Green |
| Material | Acrylic |
| For Use With (Application) | Mounting and Trim Attachment |
| Model | 8810NS |
| Quantity | 12/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.