Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotchgard™ Low Maintenance Floor Finish

Catalog No. 17623902 Shop All 3M Products
Change view
Click to view available options
Volume (English):
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
17-623-902 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-902 Supplier 3M™ Supplier No. 7010302092
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Scotchgard™ Floor Finish - Exceptionally tough and durable, low maintenance, 25% solids floor finish provides a 'wet look' gloss with less burnishing. Dries fast for outstanding image and film clarity. Use on resilient tile and terrazzo floors.

  • Use on most hard floors maintained with machines of any speed.
  • Best choice for low-to-medium traffic areas such as patient rooms, classrooms and offices
  • Use on resilient tile and terrazzo floors
  • For use with the 3M™ Easy Shine Applicator System
  • Provides a brilliant, durable, long lasting , high gloss 'wet' look
  • Easy application - low mop drag
  • Excellent leveling, dry time, and slip resistance
  • Responds quickly to spray buffing or burnishing
  • Formulation is APE-Free
  • Superior soil, abrasion, and black mark resistance compared to traditional acrylic floor finishes
TRUSTED_SUSTAINABILITY

Specifications

Product Type Floor Finish
Form Liquid
Quantity 4/Cs.
Volume (English) 1 gal.
Volume (Metric) 3.785 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.