Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ Scott™ Control Antimicrobial Foam Skin Cleanser (0.1% Benzalkonium Chloride)

Catalog No. 19413008 Shop All Kimberly-Clark Professional Products
Change view
Click to view available options
Quantity:
6 Pk.
Product Type:
Anti-bacterial Foam Soap
E-2 Foam Skin Cleanser
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
19-413-008 6 Pk. E-2 Foam Skin Cleanser
19-413-007 6 Pk. Anti-bacterial Foam Soap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-413-008 Supplier Kimberly-Clark Professional™ Supplier No. 91555
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains (0.1% Benzalkonium Chloride) to kill harmful germs such as E. Coli, Listeria, and Salmonella. Contains aloe and glycerin to moisturize the skin. Does not contain triclosan. High capacity for less refilling and maintenance time. Easy-to-load cassette clicks right into place. Bottle is recyclable after the pump is removed.

  • Scott Control E-2 Antiseptic Foam Skin Cleanser contains 1.75% PCMX
  • It is rated by the NSF as E-2 and safe for food processing and foodservice; Kills 99.9% of bacteria
  • This foaming commercial hand soap is unscented, clear and dermatologist tested
  • The foam soap comes in a high capacity container for less refilling and maintenance time
  • Easy-to-load cassette clicks right into place and is compatible with the Scott Control Professional Cassette Skin Care Dispenser

Specifications

Product Type E-2 Foam Skin Cleanser
Color Clear
Certifications/Compliance NSF rated E-2
Composition PCMX 1.75%
For Use With (Application) Kills up to 99.9% of bacteria, wash and work antiseptic formula reduces a wide range of bacteria
Quantity 6 Pk.
Volume (English) 0.26 gal.
Volume (Metric) 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.