Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ AV-3000™ Facepiece

Catalog No. 19168375
Change view
Click to view available options
Certifications/Compliance:
NIOSH, ANSI Z87.1-2003, NFPA 1981:2007
Harness Style:
Dupont Kevlar aramid fibers
Includes:
Large dual voicemitters
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Harness Style Includes
19-168-375 NIOSH, ANSI Z87.1-2003, NFPA 1981:2007 Dupont Kevlar aramid fibers Large dual voicemitters
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-168-375 Supplier Scott Safety™ Supplier No. 80577373
This item is not returnable. View return policy

Requires

To use as an air-purifying respirator, requires cartridge adapter 805622-01 for 742 Series cartridges, 805059-01 for 40mm NIOSH cartridges and 200423-01 for CBRN Cap-1 canister

Builds on the success of earlier AV-Series facepieces with design enhancements for better fit, added comfort, improved field of vision and enhanced communications

  • Top-Down Convertibility allows single facepiece for APR, PAPR, SAR and SCBA respirators
  • One fit test and reduced training
  • All with Comfort Seal with deep chin cup for reliable, comfortable fit
  • Mechanical and electronic communication options across all applications
  • All components latex free
  • Four-point Kevlar™, polyester or rubber head harness options

Order Info

Fit testing required prior to using this facepiece. Prescription lens kit (805753-01) and other accessories available separately

Certifications

All head harnesses NIOSH approved for APR, supplied air, SCBA and PAPR respirators. Rubber and Kevlar head harnesses also NIOSH approved for CBRN respirators.

Compliance

All head harnesses meet ANSI Z87.1-2003. Rubber and Kevlar head harnesses also meet NFPA 1981:2007.

Specifications

Certifications/Compliance NIOSH, ANSI Z87.1-2003, NFPA 1981:2007
Material Dupont™ Kevlar™ aramid fiber, Polyisoprene Seal
Product Type Facepiece
Format Large
For Use With (Equipment) Av-3000™ facepiece
Harness Style Dupont Kevlar aramid fibers
Includes Large dual voicemitters
Latex Free Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.