Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ AV-3000™ Facepiece with SureSeal

Catalog No. 19827928
Change view
Click to view available options
Includes:
Large dual voicemitters
Large dual voicemitters, communication bracket
Format:
Small
Medium
Large
Material:
Dupont™ Kevlar™ harness, Polyisoprene Seal
Polyester harness, Polyisoprene Seal
Rubber harness, Polyisoprene Seal
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Format Material
19-827-928 Large dual voicemitters Large Rubber harness, Polyisoprene Seal
19-500-630 Large dual voicemitters Small Dupont™ Kevlar™ harness, Polyisoprene Seal
19-500-631 Large dual voicemitters Medium Dupont™ Kevlar™ harness, Polyisoprene Seal
19-500-632 Large dual voicemitters Large Dupont™ Kevlar™ harness, Polyisoprene Seal
19-500-633 Large dual voicemitters, communication bracket Small Dupont™ Kevlar™ harness, Polyisoprene Seal
19-500-634 Large dual voicemitters, communication bracket Medium Dupont™ Kevlar™ harness, Polyisoprene Seal
19-500-635 Large dual voicemitters, communication bracket Large Dupont™ Kevlar™ harness, Polyisoprene Seal
19-827-900 Large dual voicemitters Small Polyester harness, Polyisoprene Seal
19-827-901 Large dual voicemitters Medium Polyester harness, Polyisoprene Seal
19-827-902 Large dual voicemitters Large Polyester harness, Polyisoprene Seal
19-827-926 Large dual voicemitters Small Rubber harness, Polyisoprene Seal
19-827-927 Large dual voicemitters Medium Rubber harness, Polyisoprene Seal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 19-827-928 Supplier Scott Safety™ Supplier No. 7012511866
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

To use as an air-purifying respirator, requires cartridge adapter 805622-01 for 742 Series cartridges, 805059-01 for 40mm NIOSH cartridges and 200423-01 for CBRN Cap-1 canister.

Top-Down Convertibility design allows a single facepiece for APR, PAPR, SAR and SCBA respirators.

  • One fit test and reduced training
  • Patent-pending SureSeal technology for the most advanced fit and comfort
  • Wide seal offers a more secure interface with protective suits, hoods, shrouds and helmets
  • Mechanical and extensive electronic communication options across all applications
  • All components are latex free
  • Kevlar™, polyester or rubber head harness options with three nosecup sizes

Compliance:  
All head harnesses meet ANSI Z87.1-2003. Rubber and Kevlar head harnesses also meet NFPA 1981:2007. All head harnesses NIOSH approved for APR, supplied air, SCBA and PAPR respirators. Rubber and Kevlar head harnesses also NIOSH approved for CBRN respirators.

 

Order Info

Fit testing required prior to using this facepiece. Prescription lens kit (805753-01) and other accessories available separately.

Specifications

Certifications/Compliance NIOSH, ANSI Z87.1-2003, NFPA 1981:2007
Material Rubber harness, Polyisoprene Seal
Product Type Facepiece
Format Large
For Use With (Equipment) Av-3000™ facepiece
Includes Large dual voicemitters
Latex Free Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.