Learn More
Scott Safety™ C420 PAPR Accessory, Battery Pack
Catalog No. 19818698
Change view
Click to view available options
Battery Life:
10 hr.
Catalog No. Battery Life
19-818-698 10 hr.
1 options
Catalog No. 19-818-698 Supplier Scott Safety™ Supplier No. 7012474417
  • $458.50
    $290.65 / Each of 1
    Save $167.85
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Lithium sulfur dioxide battery pack keeps PAPR charged for up to 10 hours.
  • Lithium sulfur dioxide
  • 10 hour battery life
  • 10 year shelf life
Specifications
Battery Life 10 hr.
Battery Type Lithium
Product Type Battery Pack
For Use With (Equipment) C420 Powered Air Purifying Respirator system
Model 80535801