Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ CBRN Cap-1 Canister

Catalog No. 191500863
Change view
Click to view available options
CBRN Cap-1 Canister
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-150-0863 CBRN Cap-1 Canister
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-0863 Supplier Scott Safety™ Supplier No. 7012614376

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

99.99% efficient filtration of aerosols and solid particulates for effective protection against numerous biological agents

  • Canister-type filter media is compatible with Scott full-facepiece respirators using 40mm thread connections
  • An accessory adapter allows the filter media to be used with other Scott facepieces including the Scott-O-Vista™, AV-2000™ and AV-3000™

Compliance

Meets or exceeds the C2A1 performance requirements for DMMP, Sarin (GB) and cyanogen chloride (CK).

Specifications

Certifications/Compliance NIOSH
Connection Type 40 mm NATO thread
Color Black
Material Polyamide
Product Type Cbrn Cap-1
For Use With (Application) Wide range of applications from CBRN response to riot control to hazmat response
For Use With (Equipment) M120, m112, av-2000™, av-3000™ sureseal™, c420
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.