Learn More
Scott Safety™ AV-2000 Full Facepiece Respirator Parts & Accessories: CBRN Approved 40mm First Responder Adapter
Catalog No. 19500642
Change view
Click to view available options
For Use With (Equipment):
CBRN Air-Supplied Respirators and CBRN Air-Purifying Respirators
Catalog No. For Use With (Equipment)
19-500-642 CBRN Air-Supplied Respirators and CBRN Air-Purifying Respirators
1 options
Catalog No. 19-500-642 Supplier Scott Safety™ Supplier No. 7012471464
Add to Cart
Add to Cart
Adapter allows the AV-2000 facepiece to convert from an air-supplied CBRN to an air-purifying CBRN.
Specifications
Product Type Adapter
For Use With (Equipment) CBRN Air-Supplied Respirators and CBRN Air-Purifying Respirators
Model 200423-01