Learn More
Scott Safety™ Eagle Imager™ 160 and 320Thermal Imager Replacement Battery
Catalog No. 191203886
Change view
Click to view available options
Battery Run Time:
4 hr.
Catalog No. Battery Run Time
19-120-3886 4 hr.
1 options
This item is not returnable. View return policy
Catalog No. 19-120-3886 Supplier Scott Safety™ Supplier No. 7012471382
Item Discontinued Please contact your local Fisher Scientific sales representative for pricing
This item is not returnable. View return policy
Twist-change battery for use with this high-resolution thermal imaging camera
  • One-way on, large-scale threading for fast battery replacement
  • 9V NiMH
  • Four hour run time
Specifications
Product Type Spare NiMH Battery
Battery Run Time 4 hr.
Battery Type Ni-MH
For Use With (Equipment) Eagle Imager 160/320 thermal camera
Recharge Time 2 hr.