Learn More
Scott Safety™ SCBA Accessory, EPIC™3 Voice Amplifier
Catalog No. 19100875
Change view
Click to view available options
For Use With (Equipment):
SCBA
Catalog No. For Use With (Equipment)
19-100-875 SCBA
1 options
User must verify that the EPIC 3 Voice Amplifier is approved for use on the selected Scott respirator.
Catalog No. 19-100-875 Supplier Scott Safety™ Supplier No. 7012511842
Please contact your local Fisher Scientific sales representative for pricing
User must verify that the EPIC 3 Voice Amplifier is approved for use on the selected Scott respirator.
Requires
Mounting bracket (sold separately)
Voice amplifier is an accessory for use with EPIC™3 facepiece and features automatic shut-off to reduce costs of battery replacement and improves equipment readiness.
  • Designed for use with Scott full facepieces equipped with voicemitters as part of a complete respirator
  • Amplifier electronically amplifies sound coming from the voicemitter
  • Voice amplifier assembly is equipped with a green LED light on the top of the unit to indicate powe ON as well as low battery
  • Features an automatic power OFF feature that shuts down power after 20 minutes of inactivity
  • Instrinsically safe for use with commercially available alkaline and NiMH batteries
  • Battery life of more than 50 hours is more than double that of the most popular models in the same market
  • Ribbed, textured surface establishes more secure handling even with wet-gloved hands
  • ON/OFF button with tactile surface makes it easy to recognize activation with a gloved hand
  • Angled directional speaker projects an amplified voice in the direction the use if facing
  • Upgradeable to include radio interface and talk-around, multi-channel functionality
Specifications
Certifications/Compliance NFPA, NIOSH
Applications Firefighting
For Use With (Equipment) SCBA
Model 201275-01
Weight (English) 7 oz.
Product Type Voice Amplifier