Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ Facepiece Parts

Catalog No. SCT10005135
Change view
Click to view available options
Product Type:
Facepiece Seal
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Size
SCT10005135 Facepiece Seal Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT10005135 Supplier Scott Safety™ Supplier No. 1010237680
Add to Cart
Add to Cart

Specifications

Color Black
Product Type Facepiece Seal
For Use With (Equipment) Scott-o-vista™, av-2000™ facepiece
Material Rubber
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.