Learn More
Scott Safety™ Mask Tactical Carry Bag
Catalog No. 191202841
Change view
Click to view available options
Certifications/Compliance:
NIOSH CBRN APR
Catalog No. Certifications/Compliance
19-120-2841 NIOSH CBRN APR
1 options
Catalog No. 19-120-2841 Supplier Scott Safety™ Supplier No. 7012470999
  • $206.00
    $180.65 / Each of 1
    Save $25.35
    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Tactical bag with belt mounting for use with full facepiece respirators.
Specifications
Product Type Tactical Facepiece Bag
Material Cordura™ Nylon
Model 10011882
Includes Belt Loop Attachment, Optional Leg Strap
For Use With (Equipment) M110 Dual Lens and M120 Facepiece
Certifications/Compliance NIOSH CBRN APR