Learn More
Scott Safety™ Lens Kit
Catalog No. 19168741
Change view
Click to view available options
Product Type:
Lens Kit
For Use With (Application):
High performance eye protection
For Use With (Equipment):
AV-2000 or Scott-Safety O-Vista facepieces
AV-3000 facepiece
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Application) For Use With (Equipment)
19-168-741 Lens Kit High performance eye protection AV-2000 or Scott-Safety O-Vista facepieces
19-168-740 Lens Kit High performance eye protection AV-3000 facepiece
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-168-741 Supplier Scott Safety™ Supplier No. 7012618948
Add to Cart
Add to Cart
Prescription-ready, 60mm (2.36 in. ) lens kit
  • Frame only
  • 60mm (2.36 in.) lens size
  • Stainless steel screws
  • High-performance eye wires for a secure lens fit
  • Fisher P/N 19-168-741 is for use with Scott AV-2000 (P/N 19-060-637) or Scott-O-Vista facepieces
  • Fisher P/N 19-168-74 is for use with Scott AV-3000 (P/N 19-827-900) facepiece
Specifications
Product Type Lens Kit
For Use With (Equipment) AV-2000 or Scott-Safety O-Vista facepieces
For Use With (Application) High performance eye protection
Includes Frame, stainless steel screws, 60mm lens size, high performance eye wire