Learn More
Scott Safety™ Safety Loctite™ 768™ X-NMS Cleanup Solvents
Catalog No. 19818518
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-818-518 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. 19-818-518 Supplier Scott Safety™ Supplier No. 7012473197
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used in the diassembly of parts and cleanup of dispensing equipment.
  • Dissolves cured cyanoacrylate-based adhesives
  • 1.75 oz.
Specifications
Product Type Cleanup Solvent
For Use With (Application) Used in the diassembly of parts and cleanup of dispensing equipment
Quantity 1 Ea.
Volume (English) 1.75 oz.
Volume (Metric) 51.75 mL