Learn More
Scott Safety™ Replacement Part, Seat Gasket
Catalog No. SCT5726400
Change view
Click to view available options
Product Type:
Respirator Seat Gasket
Catalog No. Product Type
SCT5726400 Respirator Seat Gasket
1 options
Catalog No. SCT5726400 Supplier Scott Safety™ Supplier No. 7012473219
Add to Cart
Add to Cart
The seat gasket helps to prevent leakage and is a significant part of the SCBA.
  • Replacement part for SCBA air system
  • Prevents air leakage
  • Seat gasket should be inspected regularly and thoroughly checked based on usage
  • Found on the high pressure hose that connects to the air cylinder
Specifications
Model 5726400
Product Type Respirator Seat Gasket