Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ SCBA Accessory, Hip Shoulder Pad Kit

Catalog No. 19822835
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-822-835 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-822-835 Supplier Scott Safety™ Supplier No. 7012473880
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fire resistant hip shoulder pad replacement part for use with Scott Safety SCBA.

Specifications

Color Black
For Use With (Equipment) SCBA
Material Dupont™ Kevlar™ aramid fiber
Model 80381004
Product Type Shoulder Pad Kit

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.