Learn More
Scott Safety™ Quantitative Fit Test Adapter
Catalog No. 19122046
Change view
Click to view available options
Includes:
Probe
Catalog No. Includes
19-122-046 Probe
1 options
Catalog No. 19-122-046 Supplier Scott Safety™ Supplier No. 7012474449
Add to Cart
Add to Cart
Ensures proper respirator fit.
  • Enables fit testing that is simple and easy to perform
  • Probed adapter assemblies permit quantitiative fit testing with all Scott half and full facepieces
  • Individuals can be fit tested in their own facepieces
  • Adapters are facepiece specific and offered for all Scott Safety facepieces
Specifications
Product Type Fit Test Adapter
For Use With (Application) Quantitative Fit Testing
Includes Probe
Model 805628-01