Learn More
Scott Safety™ SCBA Accessory, Quarter-Turn Respirator Cartridge Adapter
Catalog No. 19700874
Change view
Click to view available options
1/Each
Catalog No.
19-700-874 1/Each
1 options
This item is not returnable. View return policy
Catalog No. 19-700-874 Supplier Scott Safety™ Supplier No. 7012474448
This item is not returnable. View return policy
Adapter for use with twin series full facepiece respirators.
As part of Scott's Top-Down Convertibility, the full line of 742 Series twin cartridges can be utilized with the addition of a Quarter-Turn Adapter. Low profile, allowing for use of communications.
Specifications
Certifications/Compliance NIOSH
Applications Firefighting
For Use With (Equipment) SCBA
Model 805622-01
Quantity 1 Ea.
Product Type Adapter