Learn More
Scott Safety™ Anti-Fog Respirator Wipe
Catalog No. 19121034
Change view
Click to view available options
For Use With (Equipment):
Elsa™ emergency escape breathing respirator
Catalog No. For Use With (Equipment)
19-121-034 Elsa™ emergency escape breathing respirator
1 options
Catalog No. 19-121-034 Supplier Scott Safety™ Supplier No. 2010173
Add to Cart
Add to Cart
Anti-fog wipes made with a combination of propylene glycol and water for use with respriatory equipment.
Individually packaged. Polyester bag resists damage from chemicals and UV light. Antifog coating promotes optimum visibility
Specifications
Product Type Respirator Wipes
Model 2010173
For Use With (Equipment) Elsa™ emergency escape breathing respirator
Quantity 1 Ea.