Learn More
Scott Safety™ E-Z Flo™ Regulators for Ska-Pak Supplied-Air Respirators
Catalog No. SCT80422101
Change view
Click to view available options
Includes:
Assembly
Regulator only
Vibralert quick-disconnect
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
SCT80422101 Regulator only
19-818-602 Vibralert quick-disconnect
SCT80437001 Assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SCT80422101 Supplier Scott Safety™ Supplier No. 7100293240
Please contact your local Fisher Scientific sales representative for pricing
Designed for gloved hands, enables easy breathing, and has a two-step secure attachment.
  • Mask-mounted regulator maintains positive pressure during flows of up to 500 std. liters per minute
  • Available in a continuous hose configuration, with an optional inline quick disconnect coupling
  • The low-pressure hose equipped with a swivel attachment at the mask-mounted regulator
Specifications
Applications Confined Space
For Use With (Equipment) Ska-Pak Supplied Air Respirator
Includes Regulator only
Model 80422101
Product Type Regulator