Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ Ska-Pak™ AT Escape SCBA/Supplied-Air Respirator

Catalog No. 19121775
Change view
Click to view available options
Ska-Pak AT, Kevlar harness, 10-Minute; 3000psi composite
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-121-775 Ska-Pak AT, Kevlar harness, 10-Minute; 3000psi composite
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-121-775 Supplier Scott Safety™ Supplier No. SAR423030431101

Supplied-air respirator offers automatic, hands-free transfer capability for panic-proof protection

Type C airline respirator instantly switches from external air to cylinder air if continuous air supply is interrupted. A Vibralert™ tactile alarm warns the user when transfer occurs; a flashing electronic indicator notifies wearer of low cylinder pressure. Suitable for numerous applications in hazardous industrial, confined space, and IDLH environments.

Components

  • Universal-sized AV-2000™ facepiece with Kevlar™ head harness E-Z Flo™ regulator 5-, 10- and 15-minute portable escape cylinder (3000psig composite) E-Z Klip™ cylinder retention system for quick, easy cylinder changes Choice of nylon or Kevlar harness
Specifications
    Operates at an airline inlet pressure of 80 to 115psig Transfer to cylinder occurs when pressure drops below 50psig Great for aerial apparatus

Certifications

NIOSH approved.

Compliance

Meets NFPA airflow requirements.

Specifications

Certifications/Compliance NIOSH, NFPA
Product Type Escape Breathing Apparatus
Duration 10 min.
Pressure 80 to 115 psi
Cycle Time 10 min.
For Use With (Application) Hazardous industrial, confined space, and IDLH environments
Includes 3000 psi composite cylinder, hansen fitting, av-2000™ facepiece with kevlar headnet, e-z flo™ regulator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.