Learn More
Scott Safety™ Ska-Pak™ Plus Supplied-Air Respirator
Lightweight system with streamlined design for entry into or escape from hazardous, confined space, or Immediately Dangerous to Life and Health (IDLH) situations
Specifications
Certifications/Compliance NFPA
Product Type Supplied Air Respirator System
Duration 10 min.
For Use With (Application) For entry into or escape from hazardous, confined space, or IDLH situations
For Use With (Equipment) E-Z klip cylinder retention system
View More Specs
Products 2
Catalog Number Mfr. No. Pressure Includes Price Quantity & Availability  
Catalog Number Mfr. No. Pressure Includes Price Quantity & Availability  
19-122-014
View Documents
Scott Safety™
7012623434
2216 psig Hansen, no facepiece N/A N/A
Please contact your local Fisher Scientific sales representative for pricing
19-122-012
View Documents
Scott Safety™
SAR324060425301
4500 psig Hansen HK socket, aramid fiber facepiece Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area. N/A
Description
Description
  • Features an AV-2000™ full-facepiece with Comfort Seal™ and Kevlar™ head harness for safe, secure fit
  • Allows for interface with communication devices so contact can be maintained in emergencies
  • Comes equipped with the E-Z Klip™ cylinder retention system for quick and easy cylinder changes
  • Cat. No. 19-700-884 includes the Modulair™ Quick Disconnect regulator which allows hook-up to an existing air source such as the Scott Industrial SCBA or E-Z Airline™ supplied air respirators
Order Info
Three styles of harness are available: Kevlar for high heat or spark potential environments; standard nylon; and full-body, with NIOSH-approved integrated respirator design, which holds the cylinders and acts as a fall protection harness. Cylinder options are 5- or 10-minute 3000psig fiberglass cylinders.
Specifications
Specifications
NFPA
10 min.
E-Z klip cylinder retention system
Supplied Air Respirator System
For entry into or escape from hazardous, confined space, or IDLH situations
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.