Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ Ska-Pak™ Plus Supplied-Air Respirator

Catalog No. 19818592
Catalog No. 19-818-592 Supplier Scott Safety™ Supplier No. 7012635140

Please contact your local Fisher Scientific sales representative for pricing

missing translation for 'validMainframeMsg'

Lightweight system with streamlined design for entry into or escape from hazardous, confined space, or Immediately Dangerous to Life and Health (IDLH) situations

  • Features an AV-2000™ full-facepiece with Comfort Seal™ and Kevlar™ head harness for safe, secure fit
  • Allows for interface with communication devices so contact can be maintained in emergencies
  • Comes equipped with the E-Z Klip™ cylinder retention system for quick and easy cylinder changes
  • Cat. No. 19-700-884 includes the Modulair™ Quick Disconnect regulator which allows hook-up to an existing air source such as the Scott Industrial SCBA or E-Z Airline™ supplied air respirators

Order Info

Three styles of harness are available: Kevlar for high heat or spark potential environments; standard nylon; and full-body, with NIOSH-approved integrated respirator design, which holds the cylinders and acts as a fall protection harness. Cylinder options are 5- or 10-minute 3000psig fiberglass cylinders.

Specifications

Certifications/Compliance AS NZS 1716:2012, NIOSH
Color Black
Material PVC
Product Type Supplied Air Respirator System
For Use With (Application) Entering into or escaping from hazardous, confined space, or immediately dangerous to life and health situations
Includes Hansen/foster fitting
Material (Liner) PVC (Cadmium Free)
Model 80430501
Pressure 1.5 MPa
Length (English) 393.7 in.
Length (Metric) 10 m
Diameter (Metric) 0.9 to 1.03 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.