Learn More
Scott Safety™ Wescodyne™ Plus Cleaner Disinfectant
Iodine-based solution disinfects after 10 minutes of contact time
Supplier: Scott Safety™ 31001875
Includes: Spray attachment
Description
Broad spectrum Iodophor for use on facepieces, adapters, regulators.
- Premeasured concentrate for easy mixing—just add potable water and shake
- Spray-on application for simple disinfection
- Kills 100% of all test microorganisms
- Three-year shelf life in concentrate form
Certifications
EPA registered
Specifications
Kills 100% of all test microorganisms which provides one-step cleaning and disinfecting on AV-3000, AV-2000 and weld-o-vista facepieces, adapters and regulators | |
Spray attachment | |
10 min. | |
1 gal. |