Learn More
Glass Expansion Scott Spray Chamber for SpectroBlue or Arcos II
SDP
Catalog No. 501906960
Encompass_Preferred
Scott Spray Chamber for SpectroBlue or Arcos II (48105078). Compatible with Spectro (Ametek) SpectroBlue or Arcos II ICP-OES.This Scott style spray chamber features a double-pass baffle, which efficiently removes larger droplets, producing a smaller mean droplet size entering the plasma for better precision.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6960 Supplier Glass Expansion Supplier No. 208094053
Scott Spray Chamber for SpectroBlue or Arcos II (48105078). Compatible with Spectro (Ametek) SpectroBlue or Arcos II ICP-OES.This Scott style spray chamber features a double-pass baffle, which efficiently removes larger droplets, producing a smaller mean droplet size entering the plasma for better precision.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001