Learn More
Perkin Elmer US LLC Scott Spray Chamber and GemTip Cross-Flow II Nebulizer Quick-Change Torch Module for Optima 8x00
Catalog No. 502113145
Encompass_Preferred
This module includes: 1 Slot Quartz Torch (N0780130) GemTip Cross-Flow II Nebulizer (N0780546) Alumina Injector 2.0 mm I.D. (N0781014) Scott Spray Chamber (N0770357)
Catalog No. 50-211-3145 Supplier Perkin Elmer US LLC Supplier No. N0780606
Add to Cart
Add to Cart
Request a Quote
This module includes: 1 Slot Quartz Torch (N0780130) GemTip Cross-Flow II Nebulizer (N0780546) Alumina Injector 2.0 mm I.D. (N0781014) Scott Spray Chamber (N0770357)