Learn More
Perkin Elmer US LLC Ryton Scott Spray Chamber for Optima 2x00/4x00/5x00/7x00 DV/8x00
Catalog No. 509043782
Encompass_Preferred
The Scott spray chamber is made from corrosion-resistant material, which is inert to most mineral acids including hydrofluoric acid, aqua regia and all organic solvents normally used in ICP analyses. The Scott-type design minimizes pulsations from the peristaltic pump. The Type II, two-piece design ensures leak-free performance. The removable inner tube allows the chamber to be easily disassembled for cleaning.
Catalog No. 50-904-3782 Supplier Perkin Elmer US LLC Supplier No. N0770357
The Scott spray chamber is made from corrosion-resistant material, which is inert to most mineral acids including hydrofluoric acid, aqua regia and all organic solvents normally used in ICP analyses. The Scott-type design minimizes pulsations from the peristaltic pump. The Type II, two-piece design ensures leak-free performance. The removable inner tube allows the chamber to be easily disassembled for cleaning.