Learn More
Perkin Elmer US LLC Scott Spray Chamber Viton O-Ring for ELAN/NexION 300/350
Catalog No. 502104390
Encompass_Preferred
This O-ring is included with the Complete HF-Resistant Scott Spray Chamber (N8120124).
Catalog No. 50-210-4390 Supplier Perkin Elmer US LLC Supplier No. WE013060
This O-ring is included with the Complete HF-Resistant Scott Spray Chamber (N8120124).