Learn More
Perkin Elmer US LLC Scott/Cross-Flow Sample Introduction Torch Module for Avio 200
Supplier:  Perkin Elmer US LLC N0790606
Encompass_Preferred
This module kit includes: External Spray Chamber, Qty. 1 (N0791499) Inner Chamber for External Ryton/Type II Spray Chamber, Qty. 1 (N0582258) 2 mm I.D. Alumina Injector, Qty. 1 (N0791183) 1-Slot Quartz Torch, Qty. 1 (N0790131) End Cap Assembly for GemTip Cross-Flow Nebulizer, Qty. 1 (N0780546) Adjustable Torch Mechanical Assembly, Qty. 1 (N0790138) Century Scott/Cross-Flow II Injector Adapter Kit, Qty. 1 (N0790156) Spray Adapter, Qty. 1 (N0790296) External Drain Kit, Qty. 1 (N0790613)
Catalog No. 50-211-2371