Learn More
Leica Biosystems SCOTTS TAP WATER WORKING SOLUT
Supplier:  Leica Biosystems 3802901
Encompass_Preferred
Scotts Tap Water substitute is a bluing reagent designed for histology and cytology. It blues hematoxylin gently.Scott's Tap Water Substitute is prepared to supply a gentle but appropriate bluing solution for staining. This formulation provides bluing of nuclear chromatin and nuclear membranes.
Catalog No. 50-255-2427