Learn More
MSC Bouton Scout OTG Safety Glasses, Polycarbonate, Gray Frame, Anti-Scratch, 1 ea
Catalog No. 5000688770
Encompass
These safety glasses are designed for versatility and comfort, featuring an over-the-glass construction that allows them to be worn over most prescription eyewear or independently. Crafted from durable polycarbonate, they provide reliable eye protection for various industrial and workplace applications.
  • Over-the-glass design fits over most prescription eyewear
  • Molded nose bridge ensures a comfortable fit for most users
  • Indirect vent temples help to reduce fogging
  • Anti-scratch coating for improved lens longevity
  • Universal frame fit
  • Meets ANSI Z87.1+ and CSA Z94.3 standards
  • Certified to EN 166, EN 170, and EN 172
  • TAA compliant
Catalog No. 50-006-88770 Supplier MSC Supplier No. 92864784
These safety glasses are designed for versatility and comfort, featuring an over-the-glass construction that allows them to be worn over most prescription eyewear or independently. Crafted from durable polycarbonate, they provide reliable eye protection for various industrial and workplace applications.
  • Over-the-glass design fits over most prescription eyewear
  • Molded nose bridge ensures a comfortable fit for most users
  • Indirect vent temples help to reduce fogging
  • Anti-scratch coating for improved lens longevity
  • Universal frame fit
  • Meets ANSI Z87.1+ and CSA Z94.3 standards
  • Certified to EN 166, EN 170, and EN 172
  • TAA compliant