Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InBios International Inc. SCoV-2 Ag Detect™ Rapid Self-Test
SDP

Catalog No. 502120102
Change view
Click to view available options
No. of Reactions:
2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
502120102 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 502120102 Supplier InBios International Inc. Supplier No. CAGS2
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed for Simplicity - The InBios COVID-19 rapid test kit is incredibly simple to use. Just collect your direct nasal swab sample, place your sample in the COVID-19 test, and wait for the results. This test kit requires no mixing and can be read visually without instrumentation!

  • Sequence analysis suggests test will detect variants currently in circulation, including KP.3.1.1.
  • Fast, Easy-To-Read Results - We developed this COVID-19 home test kit to deliver fast and clear results. Within 20 minutes, the rapid COVID-19 test results can be read visually - just about the time it takes to drink your morning latte
  • FDA Emergency Use Authorized - Be ready for immediate home testing with these COVID-19 test kits. Our COVID-19 antigen test kit can be used for symptomatic or asymptomatic individuals. See package insert for complete information.
  • All-In-One Kit - This kit comes with 2 rapid tests with separately packaged swabs and pre-measured buffer tubes. The InBios antigen test kit is safe for individuals ages 2 to 13 with adult supervision and can be used for self-testing by ages 14 and above.

Specifications

Certifications/Compliance FDA EUA
Content And Storage 20°C to 30°C
CE Marker No
Clia Complexity Waived
Detectable Analytes Antigen
Detection Method Visual
Type Rapid Test
Disposable Single-use
Includes 2 tests, 2 swabs, 2 buffers
No. of Reactions 2
Sterility Non-sterile
Form Swab
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.